VWR®, Instrument trays with lid
Lieferant: VWR Collection
216-2257EA 71.5 EUR
216-2257 216-2258 216-2255 216-2256 216-2259 216-2261 216-2262
VWR®, Instrument trays with lid
Schalen/Schüsseln/Töpfe Instrumentenschalen
These instrument trays are made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Knobbed lid
  • Smooth surfaces for fast and easy cleaning
Suitable for storage or sterilisation of scissors, forceps or other dissecting instruments.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable