PETG certified clean validation bottles, Nalgene®
Lieferant: Thermo Fisher Scientific
382035-0005
216-2270EA 1612 EUR
216-2270
PETG certified clean validation bottles, Nalgene®
Flaschen Kunststoffflaschen
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Sterile
  • Clear finish
  • PETG construction
These bottles can reduce the risk of contamination.
Standards: Validated to USP 788 and <0,5 EU/ml endotoxin.