Single wafer shippers, for 5" (125 mm) wafer handling, H 22 series
Lieferant: ENTEGRIS
H22-502-0615
216-2354EA 0 EUR
216-2354
Single wafer shippers, for 5" (125 mm) wafer handling, H 22 series
Wafer-Handling-Zubehör Wafer-Versandbehälter
Conical pocket holds one wafer face down contacting only the edge of the wafer.
  • Spring retains wafer and minimises particle generation
  • Stackable for efficient use of storage space
  • Loose or lock stacking, either tray on tray or cover on tray
  • Convenient screw on covers
  • Large area on cover provides space for labelling