Closed container, Euro-Norm, Silverline
Lieferant: BEKUPLAST
9-GR 8-GR 6-GR 10-GR
216-2553EA 79.4 EUR
216-2553 216-2552 216-2551 216-2554
Closed container, Euro-Norm, Silverline
Probenbehälter Universalbehälter
Made of PPC (polypropylene copolymer), grey.
  • Stable even at high loads when stacked
  • Iimpact-resistant, unbreakable, and dimensionally stable
  • Suitable for high loads
  • 100% recyclable
  • Suitable for machine handling, automatic conveyor technology, and storage and retrieval machines
These containers are with reinforced bottom suitable for storage and transport of materials, conveyor technologies and roller track systems.