Laboratory trays
Lieferant: PHAGOGENE
60283
216-4239EA 98.7 EUR
216-4239
Laboratory trays
Schalen/Schüsseln/Töpfe Laborschalen und Laborwannen
Highly versatile trays, made of blue PP, ideal for a wide range of applications
  • Good chemical resistance
  • Good temperature resistance
  • Autoclavable