Mehrzweckgefäße mit Deckel, Nalgene®
Lieferant: Thermo Fisher Scientific
DS5300-0507 DS5300-9609 DS5300-9910
216-8262EA 113 EUR
216-8262 216-8264 216-8265
Mehrzweckgefäße mit Deckel, Nalgene®
Dosen Dosen, Universal
Polycarbonate, clear.
  • Drill, tap or apply clamps to jars to attach heating or cooling equipment
  • Very durable
  • Autoclavable
Multipurpose jars with cover, which can be used as water baths, aquaria/terraria or freeze-drying chambers.