Disperse Orange 37, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
3-[4-(2,6-Dichloro-4-nitrophenylazo)-N-ethylanilino]propionitrile
21603-5G
21603-5GEA
44.3
EUR
21603-5G
Disperse Orange 37, Sigma-Aldrich®
Disperse Orange 37
Disperse Orange 37 may be used during an experimental study conducted to understand the mutagenic and carcinogenic potential of textile azo dye processing plant effluent, that is known to have an impact on drinking water source.
Formel:
C₁₇H₁₅Cl₂N₅O₂ Molecular Weight: 392,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00181820 CAS-Nummer: 13301-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...