Methylene violet ≥65% Dye, certified_Biological Stain Commission, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
7-(dimethylamino)phenothiazin-3-one, 3H-Phenothiazin-3-one, 7-(dimethylamino)-, 7-dimethylaminophenothiazin-3-one
MQ100
216313-10G
216313-1G
216313-10GEA
676
EUR
216313-10G
216313-1G
Methylene violet ≥65% Dye, certified_Biological Stain Commission, Sigma-Aldrich®
Methylene violet
Certified for use as a constituent of MacNeal′s tetrachrome stain for blood.
Formel:
C₁₄H₁₂N₂OS Molecular Weight: 256.323 g/mol Schmelzpunkt: 216 °C (Zers., Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00005064 CAS-Nummer: 2516-05-4 EINECS: 219-733-4 |
|
Spezifikation Testergebnisse
Appearance (Colour) | Black |
Appearance (Form) | Powder |
Identity by IR | Conforms to Structure |
Wave Length | ≥253 nm |
SD UVVI E580 NM | ≥20000 |
SD UVVI E481 NM | ≥10000 |
SD UVVI E287 NM | ≥20000 |
SD UVVI E253 NM | ≥20000 |
Dye Content | ≥65% |
Biological Stain Commision, Commission Certified Number: | Pass |
Carbon | 42.60% |
Nitrogen | ≥7.100% |
TLC | Conforms |
Solubility (Colour) | Purple to Dark Purple |
Solubility (Turbidity) | Clear |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...