Methylene violet ≥65% Dye, certified_Biological Stain Commission, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 7-(dimethylamino)phenothiazin-3-one, 3H-Phenothiazin-3-one, 7-(dimethylamino)-, 7-dimethylaminophenothiazin-3-one

MQ100

216313-10G 216313-1G
216313-10GEA 676 EUR
216313-10G 216313-1G
Methylene violet ≥65% Dye, certified_Biological Stain Commission, Sigma-Aldrich®
Methylene violet

Certified for use as a constituent of MacNeal′s tetrachrome stain for blood.

Formel: C₁₄H₁₂N₂OS
Molecular Weight: 256.323 g/mol
Schmelzpunkt: 216 °C (Zers., Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00005064
CAS-Nummer: 2516-05-4
EINECS: 219-733-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7824f4316d13aa0ed6c17f6138d833adc7df1431&vid=7824f4316d13aa0ed6c17f6138d833adc7df1431&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Black
Appearance (Form) Powder
Identity by IR Conforms to Structure
Wave Length ≥253 nm
SD UVVI E580 NM ≥20000
SD UVVI E481 NM ≥10000
SD UVVI E287 NM ≥20000
SD UVVI E253 NM ≥20000
Dye Content ≥65%
Biological Stain Commision, Commission Certified Number: Pass
Carbon 42.60%
Nitrogen ≥7.100%
TLC Conforms
Solubility (Colour) Purple to Dark Purple
Solubility (Turbidity) Clear


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR