Amberlite® IRA743 freie Base Ionenaustauscherharz, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

216445-1KG 216445-250G
216445-1KGEA 391 EUR
216445-1KG 216445-250G
Amberlite® IRA743 freie Base Ionenaustauscherharz, Supelco®
Amberlite® IRA743

Amberlite® IRA743 free base is of styrenic, macroreticular form. Weakly basic resin with N-methylglucaomine functionality which is useful in boron-specific applications.


Borate-specific chelating resin for the removal of borate, boric acid, other boron species from water; highly selective so that salts, including bases, do not interfere significantly.

MDL: MFCD00132701
CAS-Nummer: 63181-98-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR