Amberlite® IRA-410 (Cl) Ionenaustauscherharz, Supelco®

Lieferant: Merck

Synonyme: Amberlite® IRA-410 Cl type

M-Clarity™ Qualitätsstufe = MQ200

216569-500G
216569-500GEA 190 EUR
216569-500G
Amberlite® IRA-410 (Cl) Ionenaustauscherharz, Supelco®
Amberlite® IRA-410 (Cl)

Amberlite® IRA-410 was used as packing material for columns. Amberlite® IRA-410 was also used in urinary analysis by HPLC.


Strongly basic (type II) anion exchange resin for water conditioning, removing iodine ions, neutralization of solutions, capturing xanthan and xanthene dyes, etc.; better regeneration efficiency than type I base resin.

MDL: MFCD00132710
CAS-Nummer: 9002-26-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR