Dibromofluorescein 95% Dye, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: 4',5'-Dibromofluorescein

216720-5G
216720-5GEA 54.3 EUR
216720-5G
Dibromofluorescein 95% Dye, Sigma-Aldrich®
Dibromofluorescein

4',5'-Dibromofluorescein, Dye content 95 % (Solvent Red 72)

Formel: C₂₀H₁₀Br₂O₅
Molecular Weight: 490.10 g/mol
Schmelzpunkt: 270…273 °C (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005042
CAS-Nummer: 596-03-2
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9ffdabeed5bbe183d65db8b57f460e718c994ed&vid=6a58cf468133b5f6d9ba0c0add8764d2e316c8b9&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR