SBW glass stoppers, standard ground joint, octagonal
Lieferant: DWK Life Sciences
246240604
217-9125EA 7 EUR
217-9125
SBW glass stoppers, standard ground joint, octagonal
Stopfen Glasstopfen
These stoppers are made from SBW glass and have an octagonal head for good grip.
  • Solid form
  • Available amber or clear
Standards: DIN 12252