Bariumcarbonat ≥99,0%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Achtung

21702.235EA 82.5 EUR
21702.235 21702.292
Bariumcarbonat ≥99,0%, AnalaR NORMAPUR® analytisches Reagens
Barium carbonate
Formel: BaCO₃
Molecular Weight: 197.34 g/mol
Schmelzpunkt: 1350 °C
Dichte: 3.89 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003448
CAS-Nummer: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1 III
Merck Index: 13,00972

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.0 %
Alkali hydroxides+carbonates(as Ba(OH)2) Max. 0.015 %
Heavy metals (as Pb) Max. 10 ppm
Insolubility in hydrochloric acid Max. 0.015 %
Oxidising substances (as NO3) Max. 50 ppm
S (Sulphur) Max. 10 ppm
Ca (Calcium) Max. 0.02 %
Cl (Chloride) Max. 20 ppm
Fe (Iron) Max. 10 ppm
K (Potassium) Max. 50 ppm
Na (Sodium) Max. 0.02 %
Sr (Strontium) Max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR