Wasserstoffperoxid 3,5% USP, J.T.Baker®

Lieferant: Avantor

2182-03EA 283.3 EUR
2182-03
Wasserstoffperoxid 3,5% USP, J.T.Baker®
Wasserstoffperoxid
Formel: H₂O₂
Siedepunkt: 100 °C (1013 hPa)
Dichte: 1,01 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011333
CAS-Nummer: 7722-84-1
EINECS: 231-765-0

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets U.S.P Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
Identification Passes Test
Acidity Passes Test
Nonvolatile Residue (USP),mg. ≤30
Barium (Ba) Passes Test
Limit of Preservative ≤0.05 %
Assay (g/100mL) 2.5 - 3.5
Particulate Matter Passes Test
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR