Tween® 20 (Polysorbat)
Lieferant: Biotium
Synonyme:
Polyoxyethylene sorbitan monolaurate
22002
22002.EA
80.2
EUR
22002.
Tween® 20 (Polysorbat)
Tween® 20 (Polysorbat)
Formel:
C₅₈H₁₁₄O₂₆ Molecular Weight: 1227,53 g/mol Dichte: 1,1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00165986 CAS-Nummer: 9005-64-5 EINECS: 500-018-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...