Decontamination bags, activated charcoal
Lieferant: Biotium
22007
22007.EA
140
EUR
22007.
Decontamination bags, activated charcoal
Elektrophorese-Reagenzien
Reagenzien zur Entfernung von Farbstoffen
Activated charcoal retains ethidium bromide, GelRed™ and GelGreen™ efficiently in a convenient bag for decontamination of used gel staining solutions. Each bag has a total capacity of at least 10 mg ethidium bromide.
- Remove mutagenic dyes (ie, ethidium bromide) from gel staining solutions
- Use to clean up any dyes from a solution to allow drain disposal
- No-mess, convenient bag
- Reusable bag capacity of at least 10 mg EtBr