Fixation buffer for microscopy and flow cytometry
Lieferant: Biotium
22015
22015.EA 173 EUR
22015.
Fixation buffer for microscopy and flow cytometry
Puffer Mikroskopie-Puffer
Ready-to-use formaldehyde-based fixation buffer for immunofluorescence staining for microscopy or flow cytometry.