Butylacetat ≥99,0%, AnalaR NORMAPUR® analytisches Reagens
Lieferant: VWR Chemicals
|
Achtung
|
22087.292EA
81.8
EUR
22087.292
22087.460
Butylacetat ≥99,0%, AnalaR NORMAPUR® analytisches Reagens
Butylacetat
Formel:
CH₃COO(CH₂)₃CH₃ Siedepunkt: 126 °C (1013 hPa) Schmelzpunkt: –76 °C Dichte: 0,875 g/cm³ (20 °C) Flammpunkt: 25 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009445 CAS-Nummer: 123-86-4 EINECS: 204-658-1 UN: 1123 ADR: 3,III Merck Index: 13,01534 |
|
Spezifikation Testergebnisse
Assay (on anhydrous substance) | Min. 99.0 % |
Acidity | Max. 0.002 meq/g |
Colouration | Max. 10 APHA |
Density (20/4) | 0.874 - 0.884 |
Evaporation residue | Max. 20 ppm |
Heavy metals (as Pb) | Max. 0.2 ppm |
Water | Max. 0.1 % |
Al (Aluminium) | Max. 0.5 ppm |
B (Boron) | Max. 0.02 ppm |
Ba (Barium) | Max. 0.1 ppm |
Ca (Calcium) | Max. 0.5 ppm |
Cd (Cadmium) | Max. 0.05 ppm |
Co (Cobalt) | Max. 0.02 ppm |
Cr (Chromium) | Max. 0.02 ppm |
Fe (Iron) | Max. 0.1 ppm |
Mg (Magnesium) | Max. 0.1 ppm |
Mn (Manganese) | Max. 0.02 ppm |
Ni (Nickel) | Max. 0.02 ppm |
Pb (Lead) | Max. 0.05 ppm |
Sn (Tin) | Max. 0.1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...