Pulvertrichter, kurzer, weiter Stiel
Lieferant: SciLabware
2130/02M
221-0002EA 297 EUR
221-0002
Pulvertrichter, kurzer, weiter Stiel
Trichter Pulvertrichter
PYREX®, borosilicate glass.
  • Chemically resistant glass
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Quick and easy to clean
Ideal for transferring powders, can also be used for liquids.