Stearinsäure 50 50, powder, GenAR® NF, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials

Synonyme: Octadecansäure

2216-05EA 170.45 EUR
2216-05
Stearinsäure 50 50, powder, GenAR® NF, Macron Fine Chemicals™
Stearinsäure

Preserve in well-closed containers. CAUTION: For Manufacturing, processing or repackaging


  • Food GMP manufactured product

Formel: C₁₈H₃₆O₂
Molecular Weight: 284.48 g/mol
Siedepunkt: 361
Schmelzpunkt: 67
Dichte: 0.94 g/cm³ (20 °C)
Flammpunkt: 196 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002752
CAS-Nummer: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product Passes Test
GMP Manufactured Product 194 - 212
Meets B.P. Chemical Specifications Passes Test
Meets E.P. Chemical Specifications ≤0.1%
Meets F.C.C. Requirements Passes Test
Meets N.F. Requirements Passes Test
Preserve in well-closed containers. ≤4.0
CAUTION: For Manufacturing, processing or repackaging 53 - 59 °C
Bulk Food Chemical 40.0 - 60.0%
Bulk Pharmaceutical Chemical ≥90.0%
NF - Identification A Passes Test
NF - Identification B (Acid Value) 194 - 212
NF - Identification C Passes Test
NF - Residue on Ignition ≤ 0.1 %
NF - Acidity Passes Test
NF - Color of Solution Passes Test
NF - Iodine Value ≤ 4.0
NF - Freezing Point 53 - 59 °C
NF - Assay (C₁₈H₃₆O₂) (stearic acid) 40.0 - 60.0 %
NF - Assay (Total of Stearic & Palmitic) ≥ 90.0 %
FCC - Congealing Temperature (solidification point - FCC) 54.5 - 69.0 °C
FCC - Iodine Value ≤ 4
FCC - Acid Value 196 - 211
FCC - Lead (Pb) ≤ 2 mg/kg
FCC - Saponification Value 197 - 212
FCC - Unsaponifiable Matter ≤ 1.5 %
FCC - Residue on Ignition ≤ 0.1 %
FCC - Water (H₂O) ≤ 0.2 %
EP/BP - Appearance Passes Test
EP/BP - Acidity Passes Test
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Nickel (Ni) ≤ 1 ppm
EP/BP - Freezing Point 53 - 59 °C
EP/BP - Iodine Value ≤ 4.0
EP/BP - Stearic Acid 40.0 - 60.0 %
EP/BP - Stearic and Palmitic Acids ≥ 90.0 %
Appearance (fine, white or yellowish-white powder)
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR