Cellulose triacetate, Selectophore™ Ionophor, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

22199-50G-F
22199-50G-FEA 291 EUR
22199-50G-F
Cellulose triacetate, Selectophore™ Ionophor, Supelco®
Cellulose triacetate
Formel: C₄₀H₅₄O₂₇
Molecular Weight: 966.845 g/mol
CAS-Nummer: 9012-09-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4932cc6a385d21d2d241a94b4fb826cf1df1713e&vid=12eafbf65ba09fb3088cae520c91859975426379&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR