Essigsäure 99.8% zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

222140010 222140025 222140200 222142500 222140050
222140010.EA 56.6 EUR
222140010. 222140025. 222140200. 222142500. 222140050.
Essigsäure 99.8% zur Analyse
Essigsäure
Formel: H₃CCOOH
Siedepunkt: 117…118 °C (760 mmHg)
Dichte: 1,048 g/cm³
Flammpunkt: 40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036152
CAS-Nummer: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Clear liquid
Water ≤0.2 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Titration with NaOH ≥99.8 %
Dilution test Passes test
Chloride (Cl) ≤1 ppm
Sulfate (SO4) ≤1 ppm
Arsenic (As) ≤0.5 ppm
Cadmium (Cd) ≤0.1 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤0.2 ppm
Nickel (Ni) ≤0.1 ppm
Heavy metals (as Pb) ≤0.5 ppm
Acetic anhydride (C4H6O3) ≤0.01 %
KMnO4-reducing substance Passes test
K2Cr2O7-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR