Koffein ≥99,0%, AnalaR NORMAPUR® analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: Methyltheobromin, 1,3,7-Trimethylxanthin, Guaranin, 1,3,7-Trimethyl-1H,3H-purin-2,6-dion, Coffein, 7-Methyltheobromin, 1,3,7-Trimethyl-2,6(1H,3H)-purindion, Thein, 3,7-Dihydro-1,3,7-trimethyl-1H-purin-2,6-dion, 2,6-Dihydroxy-1,3,7-trimethylpurin, 1,3,7-Trimethyl-3,7-dihydro-2H-purin-2,6-dion, Teein, 1,3,7-Trimethyl-3,7-dihydro-1H-purin-2,6-dion

22234.187EA 174 EUR
22234.187
Koffein ≥99,0%, AnalaR NORMAPUR® analytisches Reagens
Koffein
Formel: C₈H₁₀N₄O₂
Molecular Weight: 194,19 g/mol
Schmelzpunkt: 235…239 °C
Dichte: 1,23 g/cm³ (19 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005758
CAS-Nummer: 58-08-2
EINECS: 200-362-1
UN: 1544
ADR: 6.1,III
Merck Index: 13,01636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07e609e1c7a199176b9e5b938ef241d166d502f9&vid=1ac1145bba3574e2401439da90f37fee7fd7610b&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.0 %
Appearance White crystalline powder
Acidity Max. 0.005 meq/g
Melting point 235 - 238 °C
Substances coloured by H2SO4 Max. 60 APHA
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.1 %
Insolubility in water (± 100°C) Max. 25 ppm
Loss on drying (100°C) Max. 0.5 %
Theobromine Max. 0.2 %
Cl (Chloride) Max. 100 ppm
SO4 (Sulphate) Max. 0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR