Ethyl-N-(diphenylmethylen)glycinat 98%, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Ethyl (diphenylmethylenamino)acetate, Ethyl 2-((diphenylmethylene)amino)acetate, Ethyl-2-((diphenylmethylene)amino)acetate, Ethyl N-(diphenylmethylene)glycinate

MQ200

222542-5G 222542-25G
222542-5GEA 190 EUR
222542-5G 222542-25G
Ethyl-N-(diphenylmethylen)glycinat 98%, Sigma-Aldrich®
N-(Diphenylmethylene)glycine ethyl ester

Schiff base which can be converted to amino alcohols and amino esters.

Formel: C₁₇H₁₇NO₂
Molecular Weight: 267.32 g/mol
Schmelzpunkt: 51...53 °C (Lit.)
Flammpunkt: 110 °C (geschlossener Tiegel)
Lagertemperatur: Kühlschrank
MDL: MFCD00010590
CAS-Nummer: 69555-14-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b2b43ccfc149bd59f1200ce59d6c6a729da132f&vid=4b2b43ccfc149bd59f1200ce59d6c6a729da132f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR