Basic blue 12 85% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Nile Blue A chloride, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium chloride

M-Clarity™ Qualitätsstufe = MQ200

222550-25G 222550-5G
222550-25GEA 278 EUR
222550-25G 222550-5G
Basic blue 12 85% (Farbstoffgehalt), Sigma-Aldrich®
Nile blue chloride (Basic blue 12)

Nile blue chloride has been used to stain non-aqueous phase in fluorescence microscopy studies.


Nile blue chloride is a green colored powder and is a member of oxazine group of dyes. It changes its color from blue (pH 9,4) to purple-red (11,0). Nile blue chloride has been used to detect DNA and lipids. In addition, it also helps to identify bacterial growth. Nile blue chloride also facilitates diagnosis of disease linked with amyloid accumulation.

Formel: C₂₀H₂₀ClN₃O
Molecular Weight: 353.85 g/mol
Lagertemperatur: Room Temperature
MDL: MFCD00011936
CAS-Nummer: 2381-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a636828aaa1ca78fc29a284c5e8143a72da4a496&vid=a636828aaa1ca78fc29a284c5e8143a72da4a496&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Dark Green to Black
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Lambda Max 634 - 642 nm
Carbon Content 56.3 - 79.4%
Nitrogen Content 9.7 - 13.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR