Ammoniumcer(IV)sulfat Dihydrat ≥98,0% (RT), Puriss. p.a. ACS analytical reagent, Fluka™

Lieferant: Honeywell Chemicals

Synonyme: Cer(IV)ammoniumsulfat Dihydrat, tetra-Ammoniumcertetrakis(sulfat) Dihydrat

22269-100G 22269-500G
22269-100GEA 177.31 EUR
22269-100G 22269-500G
Ammoniumcer(IV)sulfat Dihydrat ≥98,0% (RT), Puriss. p.a. ACS analytical reagent, Fluka™
Ammoniumcer(IV)sulfat Dihydrat
Formel: (NH₄)₂Ce(SO₄)₄·2H₂O
Molecular Weight: 632.55 g/mol
Schmelzpunkt: 130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149417
CAS-Nummer: 10378-47-9
EINECS: 231-567-4

Bestellen

Spezifikation Testergebnisse

Assay (Iodometric) 98.0 - 102.0 %
Insoluble in acid Max. 0.05 %
Iron (Fe) Max. 100 ppm
Phosphate (PO₄) Max. 300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR