Drum pump sets for solvents
Lieferant: LUTZ PUMPEN
0205-401 0205-402
223-0092EA 3680 EUR
223-0092 223-0093
Drum pump sets for solvents
Pumpen Fass/Transferpumpen
Pump tube SS 41-L-SL stainless steel, drum adapter is made with PP. For easely flammable hydrocarbons such as ethanol, gasoline, butanol, iso-propanol, kerosène, methanol and petroleum.
  • Fast assembly
  • Immediately ready for action
  • Ideal for pumping and filling thin bodied liquids
  • Compact compressed air motor powerful and reliable
The explosion proof universal motor ME II 3 is ideally suitable for transferring of many thin bodied easely flammable and combustible liquids.
Pump set consisting of: 1 Motor ME II 3, 1 pump tube SS 41-L-SL stainless steel,More Product Information