Calciumchlorid Dihydrat 99,0-103,0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens

Lieferant: VWR Chemicals
Achtung

Synonyme: Calcium Chloride Dihydrate, Calcium dichloride

22317.297EA 57.9 EUR
22317.297 22317.260 22317.230 22317.320 22317.460
Calciumchlorid Dihydrat 99,0-103,0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytisches Reagens
Calciumchlorid Dihydrat
Formel: CaCl₂·2H₂O
Siedepunkt: 1600 °C (1013 hPa)
Schmelzpunkt: 176 °C
Dichte: 1,85 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149613
CAS-Nummer: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Bestellen

Spezifikation Testergebnisse

Assay 99.0 - 103.0 %
Appearance of solution Passes test Ph.Eur.
Identification A Passes test Ph.Eur.
Identification B Passes test Ph.Eur.
Solution in water Passes test
Al (Aluminium) Passes test Ph.Eur.
Acidity or alkalinity Max. 0.002 meq/g
pH (25°C; 5 %) 4.5 - 8.5
Heavy metals (as Pb) Max. 5 ppm
Insolubility in water Max. 100 ppm
Magnesium and alkali metals Max. 0.5 %
Not precipitated by (COONH4)2 (as SO4) Max. 0.2 %
Oxidising substances Max. 30 ppm
Total N (Nitrogen) Max. 20 ppm
NH4 (Ammonium) Max. 50 ppm
PO4 (Phosphate) Max. 10 ppm
SO4 (Sulphate) Max. 50 ppm
As (Arsenic) Max. 1 ppm
Ba (Barium) Max. 30 ppm
Cu (Copper) Max. 5 ppm
Fe (Iron) Max. 3 ppm
K (Potassium) Max. 100 ppm
Mg (Magnesium) Max. 100 ppm
Na (Sodium) Max. 100 ppm
Sr (Strontium) Max. 0.05 %
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR