2,2'-(2,5-Thiendiyl)bis[5-(2-methyl-2-propanyl)-1,3-benzoxazol] 99% zur Szintillation, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
2,5-Bis(5-(tert-butyl)benzo[d]oxazol-2-yl)thiophene
223999-5G
223999-5GEA
77
EUR
223999-5G
2,2'-(2,5-Thiendiyl)bis[5-(2-methyl-2-propanyl)-1,3-benzoxazol] 99% zur Szintillation, Sigma-Aldrich®
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene
suitable for scintillation
Formel:
C₂₆H₂₆N₂O₂S Molecular Weight: 430,57 g/mol Schmelzpunkt: 199…201 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005774 CAS-Nummer: 7128-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...