Calciumbis(dihydrogenorthophosphat) Monohydrat

Lieferant: VWR Chemicals

Synonyme: Calcium phosphate, monobasic, monohydrate

22403.262EA 0 EUR
22403.262
Calciumbis(dihydrogenorthophosphat) Monohydrat
Calciumbis(dihydrogenorthophosphat) Monohydrat
Formel: H₄CaO₈P₂·1H₂O
MDL: MFCD00149602
CAS-Nummer: 10031-30-8
EINECS: 231-837-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR