tri-Calciumphosphat, TECHNICAL
Lieferant: VWR Chemicals
Synonyme:
Calciumphosphat tribasisch, Calciumphosphat
22417.293EA
0
EUR
22417.293
tri-Calciumphosphat, TECHNICAL
tri-Calciumphosphat
Formel:
Ca₃(PO₄)₂ Molecular Weight: 310.18 g/mol Dichte: 3.14 g/cm³ (20 °C) |
MDL:
MFCD00015984 CAS-Nummer: 7758-87-4 EINECS: 231-840-8 Merck Index: 12,01741 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...