Calcium phosphate tribasic Ph. Eur.

Lieferant: VWR Chemicals

Synonyme: Hydroxylapatite, tri-Calcium phosphate, HAp, Calcium Hydroxide Phosphate, Calcium hydroxyphosphate, Tribasic calcium phosphate

22421.460EA 1970 EUR
22421.460 22421.290
Calcium phosphate tribasic Ph. Eur.
Calcium phosphate tribasic

Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)

Formel: Ca₅HO₁₃P₃
Molecular Weight: 502.306 g/mol
Schmelzpunkt: 1700 °C
Dichte: 3.4 g/cm³ (20 °C)
MDL: MFCD00010904
CAS-Nummer: 12167-74-7
EINECS: 235-330-6

Bestellen

Spezifikation Testergebnisse

Assay (calculated as Ca) 35,0 - 40,0 %
Identification (PO₄) Passes test
Identification (Ca) Passes test
Cl (Chloride) ≤ 0,15 %
F (Fluoride) (skip lot) ≤ 75 ppm
SO₄ (Sulphate) ≤ 0,5 %
Elemental impurities (As) ≤ 2 ppm
Elemental impurities (Pb) ≤ 1 ppm
Fe (Iron) ≤ 400 ppm
Heavy metals (as Pb) ≤ 10 ppm
Acid-insoluble matter ≤ 0,2 %
Loss on ignition (800 ± 50°C; 30 min) ≤ 8,0 %
Residual solvents Passes test
Residual metal catalysts (skip lot): .
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 2 ppm
Ni (Nickel) ≤ 1 ppm
V (Vanadium) ≤ 1 ppm
Conforms to Ph.Eur. Passes test

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR