Amberlite® IR-120 (Na) Ionenaustauscherharz, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

224359-1KG 224359-250G
224359-1KGEA 196 EUR
224359-1KG 224359-250G
Amberlite® IR-120 (Na) Ionenaustauscherharz, Supelco®
Amberlite® IR-120 (Na)

Amberlite® IR120 Na+ form may be used in separation medium for TLC, paper chromatography and LPLC.

MDL: MFCD00132707
CAS-Nummer: 9002-23-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR