Siliciumcarbid, TECHNICAL 0,500 mm
Lieferant: VWR Chemicals
36CARB
22495.292EA
142
EUR
22495.292
XXXX36CARB
Siliciumcarbid, TECHNICAL 0,500 mm
Carborundum
Formel:
SiC Siedepunkt: < 2700 °C (1013 hPa) Schmelzpunkt: 2700 °C Dichte: 3,23 g/cm³ (20 °C) |
CAS-Nummer:
409-21-2 EINECS: 206-991-8 Merck Index: 12,08636 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...