Sodium metatungstate monohydrate

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Sodium polytungstate monohydrate, Sodium tungstate (meta) monohydrate, hexa-Sodium tungstate (W12) monohydrate

22534.18 22534.30 22534.56
22534.18EA 81.5 EUR
22534.18 22534.30 22534.56
Sodium metatungstate monohydrate
Sodium metatungstate monohydrate

CAS No.: 12141-67-2 Molecular Formula: Na6O39W12.H2O Formula Weight: 2986.13 (2968.12anhy) Physical Form: Powder Density: 5.47 Appearance: White Odour: Odourless MDL No.: MFCD00061466

Formel: Na₆O₃₉W₁₂·H₂O
Dichte: 5,47
Lagertemperatur: Raumtemperatur
MDL: MFCD00061466
CAS-Nummer: 12141-67-2
EINECS: 412-770-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR