Lanthan(III)chlorid Heptahydrat 64,5 - 70,0% (durch Trockensubstanz), BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

2255-05EA 296.2 EUR
2255-05
Lanthan(III)chlorid Heptahydrat 64,5 - 70,0% (durch Trockensubstanz), BAKER ANALYZED® ACS, J.T.Baker®
Lanthan(III)chlorid Heptahydrat

For flame enhancement in atomic absorption spectroscopy.

Formel: LaCl₃·7H₂O
Siedepunkt: 860 °C (1013 hPa)
Schmelzpunkt: 857 °C
Dichte: 3,8773 g/cm³ (–0.15 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149756
CAS-Nummer: 10025-84-0
EINECS: 233-237-5
Merck Index: 13,05379

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 64.5 - 70.0%
For Laboratory,Research,or Manufacturing Use ≤0.01%
ACS - Assay (LaCl₃) 64.5 - 70.0 %
ACS - Insoluble Matter ≤0.01 %
ACS - Calcium (Ca) ≤0.001 %
Trace Impurities - Magnesium (Mg) ≤10 ppm
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR