N-Acetylmuramic acid
Lieferant: Thermo Scientific
Synonyme:
2-Acetamido-2-deoxy-3-O-(d-1-carboxyethyl)-D-glucopyranose, (R)-2-(((2R,3R,4R,5R)-2-Acetamido-4,5,6-trihydroxy-1-oxohexan-3-yl)oxy)propanoic acid
226632500
226632500.EA
272
EUR
226632500.
N-Acetylmuramic acid
N-Acetylmuramic acid
Formel:
C₁₁H₁₉NO₈ Molecular Weight: 293,27 g/mol Schmelzpunkt: +64° (66°C, C=0,1% H2O) Lagertemperatur: Kühlschrank |
MDL:
MFCD00221511 CAS-Nummer: 10597-89-4 EINECS: 234-214-2 Merck Index: 13,06328 |
Spezifikation Testergebnisse
Appearance | white powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Melting point | 124°C to 132°C |
Solubility | (5% in water) clear solution |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...