N-Acetyl-5-hydroxytryptamin 98%
Lieferant: Thermo Scientific
Synonyme:
N-(2-(5-Hydroxy-1H-indol-3-yl)ethyl)acetamide, N-Acetylserotonin
226930010
226930010.EA
346
EUR
226930010.
N-Acetyl-5-hydroxytryptamin 98%
N-Acetyl-5-hydroxytryptamin
Formel:
C₁₂H₁₄N₂O₂ Molecular Weight: 218,3 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00005656 CAS-Nummer: 1210-83-9 EINECS: 214-916-5 |
Spezifikation Testergebnisse
Appearance | white to light yellow powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Melting point | 119°C to 123°C |
Water | ≤1 % (K.F.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...