Chloroform ≥99% stabilisiert, GPR RECTAPUR®
Lieferant: VWR Chemicals
22707.295EA
46.7
EUR
22707.295
22707.364
22707.466
22707.320
22707.550
Chloroform ≥99% stabilisiert, GPR RECTAPUR®
Chloroform
Spezifikation Testergebnisse
Assay | Min. 99 % |
Boiling point | 59.5 - 61.5 °C |
Density (20/4) | 1.478 - 1.489 |
Evaporation residue | Max. 10 ppm |
Free chlorine | Max. 10 ppm |
Heavy metals (as Pb) | Max. 10 ppm |
Water | Max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...