Molybdenum(VI) fluoride

Lieferant: Thermo Scientific
Gefahr

22713.18
22713.18EA 2320 EUR
22713.18
Molybdenum(VI) fluoride
Molybdenum(VI) fluoride

CAS No.: 7783-77-9 Molecular Formula: F6Mo Formula Weight: 209.93 Physical Form: Liquid, Packed in one-way cylinder w/CGA 330 valve Density: 2.3 Sensitivity: Moisture Sensitive Appearance: Colourless Melting Point: 17° Boiling Point: 37° UN No.: UN3289 MDL No.: MFCD00014220

Formel: F₆Mo
Molecular Weight: 209,94 g/mol
Siedepunkt: 37 °C
Schmelzpunkt: 17 °C
Dichte: 2,3
Lagertemperatur: Raumtemperatur
MDL: MFCD00014220
CAS-Nummer: 7783-77-9
EINECS: 232-026-5
ADR: 6.1,II
Merck Index: 14,6237

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR