Ethanolamin, Wasser_0,5 99
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Aminoethanol, Monoethanolamin
22793.30
22793.30EA
51
EUR
22793.30
Ethanolamin, Wasser_0,5 99
Ethanolamin
Purity: 99%
CAS No.: 141-43-5
Molecular Formula: HOCH2CH2NH2
Formula Weight: 61.08
Flash Point: 93°(199°F)
Physical Form: Liquid
Density: 1.012
Refractive Index: 1.4540
Sensitivity: Air Sensitive and Hygroscopic
Odour: Ammonia-like
Melting Point: 10°
Boiling Point: 169-170°
Beilstein Reference: 505944
Merck Reference: 14,3727
UN No.: 2491
MDL No.: MFCD00008183
Formel:
NH₂CH₂CH₂OH Molecular Weight: 61.08 g/mol Siedepunkt: 169…170 °C Schmelzpunkt: 10 °C Dichte: 1.012 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008183 CAS-Nummer: 141-43-5 EINECS: 205-483-3 UN: 2491 ADR: 8 III Merck Index: 14,3727 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...