Cibacron Brilliant Yellow 3G-P, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Reactive yellow 2

228478-25G
228478-25GEA 283 EUR
228478-25G
Cibacron Brilliant Yellow 3G-P, Sigma-Aldrich®
Cibacron Brilliant Yellow 3G-P

Cibacron Brilliant Yellow 3G-P, (Reactive yellow 2)

Formel: C₂₅H₁₉Cl₃N₉NaOS₃
Molecular Weight: 831,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009778
CAS-Nummer: 50662-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10b72cf921ee487a71388ee431bc2af240d80962&vid=905101cb5101f747ca02ccde87bddbb67de8801c&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR