Kristallviolett 90+% ACS
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
22866.14
22866.22
22866.14EA
45.3
EUR
22866.14
22866.22
Kristallviolett 90+% ACS
Kristallviolett
Purity: 90+%
CAS No.: 548-62-9
Molecular Formula: C25H30ClN3
Formula Weight: 407.99
Physical Form: Powder
Melting Point: ca 215° dec.
Beilstein Reference: 4077708
Merck Reference: 14,4395
UN No.: 3077
MDL No.: MFCD00011750
Formel:
C₂₅H₃₀ClN₃ Dichte: 1,19 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011750 CAS-Nummer: 548-62-9 EINECS: 208-953-6 UN: 3077 ADR: 9,III Merck Index: 14,4395 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...