Citronensäure Monohydrat ≥99,5%
Lieferant: Thermo Fisher Scientific
22869.22
22869.36
22869.A1
22869.22EA
27.4
EUR
22869.22
22869.36
22869.A1
Citronensäure Monohydrat ≥99,5%
Citronensäure Monohydrat
Purity: 99.5+%
CAS No.: 5949-29-1
Molecular Formula: C6H8O7.H2O
Formula Weight: 210.14 (192.13anhy)
Physical Form: Granular
Density: 1.542
Appearance: White
Odour: Odourless
Merck Reference: 14,2326
MDL No.: MFCD00149972
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...