Ethyl violet, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Basic Violet 4, N-(4-(Bis(4-(diethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium chloride
228842-25G
228842-25GEA
115
EUR
228842-25G
Ethyl violet, Sigma-Aldrich®
Ethyl violet
Ethyl Violet,cationic triarylmethane dye (Basic violet 4; Ethyl purple 6B)
Formel:
C₃₁H₄₂ClN₃ Molecular Weight: 492,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011765 CAS-Nummer: 2390-59-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...