Tubing clamps, coated
Lieferant: HAMMACHER KARL
HSE904-20
229-0374EA 151 EUR
229-0374
Tubing clamps, coated
Klemmen Schlauchklemmen und Sanitärklemmen
Tubing clamps made of stainless steel.
  • Fingers are covered with plastic coating to protect clamped items
  • Corrosion resistant