Ersatz-Befüllhahn für Ballonflaschen
Lieferant: SEMADENI
3100
229-7489EA 90.1 EUR
229-7489
Ersatz-Befüllhahn für Ballonflaschen
Ventile Kugelventile
½” universal valve made from PP, resistant to acids, alkalis and solvents.
  • Fully sealed design, runs very easily and with no blockages
  • Outlet spout Ø 12 mm
Housing and ball made from PP, ball seal made from PTFE and O ring made from nitrile rubber (NBR 70).