o-Xylol 96 HPLC
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,2-Dimethylbenzol
22902.K2
22902.M1
22902.K2EA
113
EUR
22902.K2
22902.M1
o-Xylol 96 HPLC
o-Xylol
Purity: 96% min
CAS No.: 95-47-6
Molecular Formula: C8H10
Formula Weight: 106.17
Flash Point: 32°(89°F)
Physical Form: liquid
Density: 0.877
Refractive Index: 1.5050
Appearance: Colourless
Melting Point: -25°
Boiling Point: 143-145°
Beilstein Reference: 1815558
Merck Reference: 14,10081
UN No.: 1307
MDL No.: MFCD00008519
Formel:
C₈H₁₀ Molecular Weight: 106.17 g/mol Siedepunkt: 143…145 °C Schmelzpunkt: –25 °C Dichte: 0.877 Flammpunkt: 32 °C (89 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008519 CAS-Nummer: 95-47-6 EINECS: 202-422-2 UN: 1307 ADR: 3 III Merck Index: 14,10081 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...