Wasser, ultrapure HPLC
Lieferant: Thermo Fisher Scientific
Synonyme:
Wasserstoffoxid, Hydrogenoxid
22934.K2
22934.K7
22934.M6
22934.K2EA
53.3
EUR
22934.K2
22934.K7
22934.M6
Wasser, ultrapure HPLC
Wasser
CAS No.: 7732-18-5
Molecular Formula: H2O
Formula Weight: 18.02
Physical Form: Liquid
Density: 1.000
Refractive Index: 1.3330
Odour: Odourless
Melting Point: 0°
Boiling Point: 100°
Merck Reference: 14,10039
MDL No.: MFCD00011332
Formel:
H₂O Molecular Weight: 18.02 g/mol Siedepunkt: 100 °C Schmelzpunkt: 0 Dichte: 1.000 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 14,10039 |
Spezifikation Testergebnisse
Appearance (Color) | Clear colorless |
Form | Liquid |
Assay (unspecified) | Specific Conductance at 25°C and time packaging: 1.5 microS/cm, max |
Optical Absorbance at | 400 nm 0.005 AU, max- at 300 nm 0.005 AU, max- at 250 nm 0.005 AU, max- at 200 nm 0.010 AU, max- at 190 nm 0.010 AU, max |
Residue on Evaporation | 2 ppm, Max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...